Learn More
Trajan Scientific and Medical Gas Filter - Oxygen
Prevent contaminants in gases from significantly impacting the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation.
Brand: Trajan Scientific and Medical 1035210
Additional Details : Weight : 2.00000kg
Description
- Capacity of 150 mL oxygen for long filter lifetime
- Indicator color change tells you when to change your filter
- Can be used in combination with other filters through a simple connecting unit
Specifications
150 mL O2 | |
Oxygen | |
Connecting unit |
From green to gray | |
Removes oxygen as well as traces of sulfur and chlorine from carrier gas |
For research use only