missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Gasket For Grinding Jar
Brand: RETSCH 22.085.0005
Code : 50
Additional Details : Weight : 0.02000kg
Specifications
For 35mL grinding jar | |
10 Pack | |
Gasket |
35 mL Grinding Jar, Stainless Steel | |
Stainless Steel |