missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GC Recombinant Rabbit Monoclonal Antibody (K01_2Q40)

Product Code. 30830396
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830396 50 μL
30824768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830396 Supplier Invitrogen™ Supplier No. MA565724

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: AEDITNILSK C.

GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GC
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2Q40
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene GC
Gene Accession No. P02774, P04276, P21614
Gene Alias DBP; DBP/ VDBG; DBP/GC; DBP02; DBP-maf; epididymis secretory protein Li 51; Gc; Gc globulin; Gc protein-derived macrophage activating factor; GC, vitamin D binding protein; gc-globulin; GcMAF; Gc-MAF; GRD3; group specific component; Group specific component vitamin D binding protein; Group-specific component; group-specific component (vitamin D binding protein); hDBP; HEL-S-51; V VDB; VDB; VDBG; Vdbp; Vitamin D binding alpha globulin; vitamin D-binding alpha-globulin; vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor
Gene Symbols GC
Host Species Rabbit
Immunogen A synthetic peptide of human Vitamin D Binding protein (aa 255-265).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14473, 24384, 2638
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.