missing translation for 'onlineSavingsMsg'
Learn More

GCK 293T Cell Overexpression Lysate 2 (Denatured), Abnova

Brand:  Abnova H00002645-T02.100uL

Additional Details : Weight : 0.00010kg

Product Code. 16178604

  • 209.00 € / 100µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Western Blotting

Specifications

Specifications

Western Blot
GCK 293T Cell Transient Overexpression Lysate(Denatured)
51.26
pCMV-GCK full-length
100 μL
Human
293T
BC001890
6.8
Transient overexpression cell Lysate was tested with Anti-GCK antibody (H00002645-B02) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers