Learn More
Cytiva Accessories and Replacement Parts for Amersham™ VacuGene XL Vacuum Blotting System
VacuGene XL vacuum blotting system is the choice for nucleic acid blotting.
81.48 € - 230.52 €
Specifications
For Use With (Equipment) | Amersham™ VacuGene™ XL Vacuum Blotting System |
---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
10488824
|
Cytiva
18-1010-43 |
Gel Supporting Screen |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10321275
|
Cytiva
18-1010-44 |
Masks |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Rapidly transfer DNA from agarose gels up to 20 x 30cm in less than 1 hour
- Efficient and reproducible transfers improve blotting results
- Carry out all pretransfer steps in the unit
- Precise vacuum control for optimal transfer conditions
- RNA transfers in less than 4 hours
Specifications
Amersham™ VacuGene™ XL Vacuum Blotting System |