missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Blot Module for miniVE Vertical Electrophoresis System
Performs 1-D and 2-D electrophoresis and electrotransfer in a single compact unit
Brand: Cytiva 80-6418-96
Code : GE
Additional Details : Weight : 0.81200kg
Includes: 3 Dacron sponges 6mm (0.25 in. thick), 25 sheets of blotter paper
Description
- Can Cast, run, and blot mini-gels in four streamlined components: gel module, blot module, lower buffer chamber, safety lid
- Can run two mini-gels or four mini-blots at once, using 10 × 10.5cm gel cassettes for increased separation distances
- Complete semi-wet blotting in 45min with only 300ml buffer
- Precast gels can be used from a variety of manufacturers
- Perform two second-dimension separations using 7cm IPG strips in 1.5 hour
- Power requirement 15w per blot module
- Working temperature is 5° to 75°C
Specifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE | |
Blot Module | |
41°F to 167°F | |
4.1 in. | |
3.93 in. | |
15 w |
MiniVE™ Vertical Electrophoresis System | |
3 Dacron sponges 6mm (0.25 in. thick), 25 sheets of blotter Paper, supplied with blot module | |
5°C to 75°C | |
10.5 cm | |
10 cm |