Learn More
Cytiva Equilibration Tubes
Provides a convenient means for equilibrating IPG strips prior to 2-dimension electrophoresis
Brand:  Cytiva 80-6467-79
Code : GE
Additional Details : Weight : 0.50000kg
Product Code. 11713773
  • 77.39 € / 12 sets
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Includes: Recommendation regarding equilibration solution volumes
Description
Description
  • Complete packsize: 12
Specifications
Specifications
For equilibrating IPG strips prior to 2-dimension electrophoresis