missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelatin Type A Type A; 225 Bloom Food Grade MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
Quantity | 100 g |
Grade | Food |
Chemical Name or Material | Gelatin Type A, Type A; 225 Bloom |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'