Learn More
Fisherbrand™ General Purpose Digital Heating Circulator
This general purpose digital heated circulator provides great temperature stability and control for demanding applications requiring accurate temperature control.
959.30 € - 1102.40 €
Product Code | Brand | Temperature Range | External Circulation | Weight (Metric) | Voltage | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range | External Circulation | Weight (Metric) | Voltage | Price | Quantity & Availability | ||
17265556
|
Fisherbrand™
|
0°C to 100°C | No | 2.1 kg | 120/230 V |
959.30 €
Each |
In Stock
Log in to see stock. |
||
17225566
|
Fisherbrand™
|
-20°C to +120°C | Yes | 2.3 kg | 230 V |
1102.40 €
Each |
In Stock
Log in to see stock. |
||
Description
- Temperature ranges from ambient 5°C to 100°C (using baths larger than 5 L), to ambient 15°C to 100°C (with 5 L baths)
- Cooling/heating capacities can be reached with a cooling unit
- Simple, intuitive user interface with dial and two function buttons
- Clear, bright 4-digit LED display
- Visual alarm and low liquid detection
- 2-point user calibration
- 3 temperature presets
Applications:
- Clinical, Microbiology and Pathology labs for media tempering, thawing, and incubating samples
- University research for temperature control of spectrophotometers, refractometers and jacketed vessels
- Industrial labs for temperature probe calibration, water analysis, QC testing product, petrochemical testing, material testing, and milk sample testing
Specifications
Digital | |
4-digit LED | |
50/60 Hz |