missing translation for 'onlineSavingsMsg'
Learn More

S Murray™ General-Purpose Narrow-Neck Bottles

Product Code. 12811304
Click to view available options
Capacity (Metric):
10,000 mL
5000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Use these bottles for general storage. S Murray™ General-Purpose Narrow-Neck Bottles feature molded graduations and s covered swivel handle.

  • High density polyethylene, octagonal bottles
  • With screw cap

Specifications

Capacity (Metric) 5000 mL
Color Natural
For Use With (Application) For storage and transportation of liquids
Material MDPE
Closure Included Yes
Closure Type Screw Cap
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'