Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Brand:  Thermo Scientific™ 840-277800
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15550592
  • 207.50 € / Each
Estimated Shipment: 29-05-2024
to see stock.
Description
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
Specifications
Specifications
This sample accessory holds one cylindrical long path cell holder with 5-100 mm path length. Comes assembled in a removable sample compartment liner.
Measurements of gases and select older standard methods that call specifically for cylindrical cells
5 to 100 mm cylindrical cuvette (1)
Long Path Cell Holder
GENESYS 30/40/50 Spectrophotometers
Cylindrical