missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin sulfate solution-10, MP Biomedicals™

Product Code. p-4404600 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mL
5 x 10 mL
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 11491822

Brand: MP Biomedicals 0210503010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1405-41-0
MDL Number MFCD00270181
Synonym Garamycin, Genoptic

Specifications

Type Gentamicin Sulfate
Physical Form Liquid
Color Colorless to Yellow
Quantity 10 mL
Grade USP
Chemical Name or Material Gentamycin Sulfate Solution - 10, Cell Biology and Analysis
compliance-icons compliance-icons
Product Identifier
  • Gentamicin Sulfate Solution-10
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Gentamicin Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.