Learn More
GGCT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556295
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NSGCKDVTGP DEESFLYFAY GSNLLTERIH LRNPSAAFFC VARLQDFKLD FGNSQGKTSQ TWHGGIATIF QSPGDEVWGV VWKMNKSNLN SLDE Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 83%.
GGCT Polyclonal antibody specifically detects GGCT in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
GGCT | |
Polyclonal | |
Unconjugated | |
GGCT | |
A030007L17Rik; AA673177; C7orf24; CRF21; Cytochrome c-releasing factor 21; gamma -glutamyl cyclotransferase; gamma-glutamyl cyclotransferase; gamma-glutamyl cyclotransferase-like 1; gamma-glutamylcyclotransferase; Gctg; GGC; GGCT; Ggctl1; RGD1304876; RGD1562590 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79017 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75223 | |
GGCT | |
Recombinant protein corresponding to Human GGCT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only