missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ GHS Symbol - Corrosive, Individual Markers on a Roll
Pre-printed labels to identify your chemical product containers.
Brand: Brady™ 834181
74.76 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Additional Details : Weight : 0.04320kg
Description
- High-quality labels that can withstand the most demanding environments
- Highly resistant to bad weather, abrasion and chemical products
Specifications
Gloss | |
250/Pk. | |
21 mm | |
21 mm | |
Diamond |
Polyester | |
Permanent Acrylic | |
0.099 mm | |
-40°C to 110°C | |
Black on White |