Learn More
Gibco™ DMEM/F-12, powder
Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells.
134.00 €
Specifications
Content And Storage | Storage conditions: 2-8° C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
---|---|
Form | Powder |
Serum Level | Standard Serum Supplementation |
With Additives | High Glucose, Glutamine, Phenol Red, Sodium Pyruvate |
Without Additives | No HEPES |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17122949
|
Gibco™
12500096 |
50 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM / F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM / F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM / F-12 is modified as follows:
With: L-glutamine, Phenol Red
Without: HEPES, Sodium Bicarbonate
The complete formulation is available.
Gibco™ DMEM / F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids and vitamins with F-12's wide variety of components.
Using DMEM/F-12
DMEM / F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2-8° C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
|
Standard Serum Supplementation | |
No HEPES | |
Gibco™ |
Powder | |
High Glucose, Glutamine, Phenol Red, Sodium Pyruvate | |
cGMP-compliant under the ISO 13485 standard | |
Room Temperature |