missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Horse Serum, New Zealand origin
Excellent value for basic cell culture, specialty research, and specific assays
29.93 € - 118.00 €
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Adult |
Species | Horse/Equine, Horse |
Country of Origin | New Zealand |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11520516
|
Gibco™
16050130 |
100 mL |
29.93 €
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11510516
|
Gibco™
16050122 |
500 mL |
118.00 €
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays
- It has earned the trust of researchers with consistent quality and award-winning support
- Helps meet research needs and budget requirements
- EIA tested (negative) serum from a donor herd
- Shipped frozen, store at <10°C
- Sera Category: Specialty
- Origin: New Zealand
Order Info
Shipping Condition: Dry ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Horse/Equine, Horse | |
Liquid | |
Standard (Sterile-filtered) | |
Gibco™ |
Adult | |
New Zealand | |
Horse Serum | |
Sustainable packaging | |
Frozen |
For In Vitro Diagnostic Use.