missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-SHH, Clone: 8G3, Abnova™
Brand: Abnova MAB10317.100uL
Additional Details : Weight : 0.02000kg
Specifications
SHH | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
SHH | |
SHH | |
Recombinant protein corresponding to human SHH. | |
RUO | |
6469 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Western Blot | |
8G3 | |
Mouse monoclonal antibody raised against recombinant SHH. | |
In ascites (0.03% sodium azide) | |
HHG1/HLP3/HPE3/MCOPCB5/SMMCI/TPT/TPTPS | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |