Learn More
Gibco™ PB-MAX™ Karyotyping Medium
Fully supplemented medium developed for short-term culture of peripheral blood lymphocytes for cytogenetic studies and in vitro diagnostic procedures
67.25 € - 278.00 €
Specifications
Additive | L-Glutamine |
---|---|
Color | Red |
Content And Storage | Storage conditions: -5 to -20°C. Protect from light. Shipping conditions: Frozen Shelf life: 12 months from date of manufacture |
Dilution | 1 X |
Product Line | PB-MAX™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10722057
|
Gibco™
12557013 |
100 mL |
67.25 €
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11589716
|
Gibco™
12557021 |
500 mL |
278.00 €
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Ready-to-use format
- Quality and performance testing
- Classic formulation
- Sustainable packaging
1X medium is a frozen, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2° to 8°C for up to 14 days.
Quality and performance testing
Every lot of is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Peripheral blood lymphocytes from a normal adult donor are cultured for 72 hours in Gibco PB-MAX Karyotyping Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot is tested for sterility, pH, osmolality, and mycoplasma.
Classic formulation
Gibco PB-MAX Karyotyping Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), L-glutamine, and phytohemagglutinin (PHA). Gibco PB-MAX Karyotyping Medium is based on formulations referenced in the Association for Cytogenetic Technologists Laboratory Manual for the culture of peripheral blood lymphocytes for cytogenetic analysis.
Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Karyotyping, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Compliance
Manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
L-Glutamine | |
Storage conditions: -5 to -20°C. Protect from light. Shipping conditions: Frozen Shelf life: 12 months from date of manufacture |
|
PB-MAX™ | |
Human | |
Sustainable packaging | |
Standard Serum | |
Sterile-filtered |
Red | |
1 X | |
Karyotyping Medium | |
Cell Viability; Proliferation & Function; Clinical & Translational Research; Mammalian Cell Culture | |
Lymphocytes | |
Dry Ice | |
Liquid |
For in vitro diagnostic use.