missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse Sonic Hedgehog C25II Recombinant Protein
Recombinant Sonic Hedgehog C25II is a bioactive protein intended for use in cell culture applications
333.00 € - 11240.00 €
Specifications
Accession Number | BC063088 |
---|---|
For Use With (Application) | Cell Culture |
Gene ID (Entrez) | 6516263 |
Molecular Weight (g/mol) | 19.7 kDa |
Purification Method | Sequential Chromatography |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10533674
|
Gibco™
PMC8034 |
10μg |
333.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10398143
|
Gibco™
PMC8035 |
25μg |
616.00 €
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10164903
|
Gibco™
PMC8031 |
100μg |
1886.00 €
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10053163
|
Gibco™
PMC8033 |
1mg |
11240.00 €
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mouse Sonic Hedgehog C25II Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Sonic Hedgehog is instrumental in patterning the early embryo. It has been implicated as the key inductive signal in patterning of the ventral neural tube, the anterior-posterior limb axis, and the ventral somites.Shipping Condition: Wet Ice.
Specifications
BC063088 | |
6516263 | |
Sequential Chromatography | |
SHH | |
Mouse | |
Carrier-Free | |
Wet Ice | |
E. coli | |
Recombinant, Ligand, Mutant | |
Sonic Hedgehog | |
Gibco™ |
Cell Culture | |
19.7 kDa | |
Sonic Hedgehog C25II | |
175 aa | |
This product is supplied lyophilized. Store at 2 to 8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤ -20°C. Avoid repeated freeze⁄thaw cycles. | |
< 0.1 ng/μg | |
5×103 to 0.83 × 102 units⁄mg | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Signal Transduction, Development, Differentiation, Stem Cell Research, Oncology | |
95 % |
For Research Use Only. Not for use in diagnostic procedures.