missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ Accessory for PIPETMAN™ Complete Pipetting System - Reagent Reservoir
Ideal for dispensing reagents with multichannel pipettes. Gilson™ Accessory for PIPETMAN™ Complete Pipetting System- Reagent Reservoirs' unique shape allows for efficient and nearly complete sample withdrawal.
208.00 € - 335.00 €
Specifications
Color | White |
---|---|
For Use With | All Pipettes with Tips Mounted |
Material | Polystyrene |
For Use With (Equipment) | PIPETMAN™ Complete Pipetting System |
Quantity | 100 Pk. |
Description
- Angled sides and V-shaped bottoms
- Pour-off spouts on all corners
- Graduations on inside walls at middle volume
Specifications
White | |
Polystyrene | |
100 Pk. |
All Pipettes with Tips Mounted | |
PIPETMAN™ Complete Pipetting System |