Learn More
Gilson™ MICROMAN™ Positive-Displacement Pipets
Ensures accuracy and precision when pipetting problem liquids, and offers full protection against cross contamination. Gilson™ MICROMAN™ Positive-Displacement Pipets are 3 to 4 times better than an air displacement pipet when pipetting viscous or volatile liquids.
Brand: Gilson™ F148505
Code : A0
Additional Details : Weight : 1.00000kg
Description
- Piston is located inside the tip or capillary preventing pipet contamination
- Color-coded push-button allows for easy identification of different models
- Dual function push-button that also ejects capillary pistons
- Comfortable PVDF handle resistant to aggressive chemicals
- Accurately measure high vapor pressure liquids without leakage or degradation of the tip
- M10 and M100 models are fully autoclavable
Specifications
C250 | |
M250 | |
Positive Displacement Pipette | |
Pipetting of viscous and dense liquids |
Autoclavable | |
1 | |
50 to 250 μL |