Learn More
Gilson™ PIPETMAN Neo™ Multichannel Pipets
Offers an excellent compromise between comfort and target precision. Gilson™ PIPETMAN Neo™ Multichannel Pipets are designed to apply equal pressure on each piston to ensure consistent volume levels across all channels.
Brand: Gilson™ F14403
Code : NEW
Additional Details : Weight : 0.32400kg
Description
- Easy-to-remove tip ejector with autoclavable lower part
- Internal design applies equal pressure on each piston to ensure consistent volume levels across all channels
- PVDF handle minimizes the hand-warming effect during pipetting and is resistant to aggressive chemicals
- Gilson Trident and unique spacer create a stepped ejection, staggering the tip ejection and thus reducing the force required to eject tips
- Unique tip holder allows for a secure fit of most brands of tips
- Available in 8- and 12-channel models
Specifications
Autoclavable | |
8 | |
Multi-Channel Pipette | |
1 Year |
P8x200N | |
Pipetman™ Neo™ | |
20 to 200 μL |