missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Stirred Bottle Reactor, PP / PTFE / PEEK / Stainless Steel Contact surfaces
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Description
Self contained bottle stirring system ideal for simple mixing processes. Stirrer shaft can be adjusted to the optimal position in either a 500 or 1000 mL DURAN™ GL 45 bottles. Use with standard magnetic stirrers up to 500 rpm. Significantly improved mixing performance compared to standard magnetic stir bars. Two GL 14 port connector cap is compatible with the modular DURAN™ connection systems; tubing with 1.6 mm to 6.0 mm bores can be used to add or remove liquid or gas. Fully autoclavable. Parts in contact with media conform to FDA requirements. Supplied with or without bottle.
- The variable stirrer shaft can be adjusted to the optimal position in either 500 or 1000 ml DURAN™ GL 45 bottles
- Compatible with 1.6 to 6.0 mm outer diameter semi-rigid tubing for addition or removal of liquids or gas
- Use with standard magnetic stirrers up to 500 rpm and allows a clearly improved mixing
- Fully compatible with the DURAN™ connection systems
Specifications
Capacity (Metric) | 500 mL |
Description | GL 45 stirred reactor, incl. 500 mL DURAN GL 45 Bottles, folding magnetic stirrer and GL 45 pp cap with 2 x GL 14 ports, 2 x GL 14 PBT caps red |
For Use With (Application) | Simple, low cost, bioreactor; Simple continuous flow stirred reactor; Mixing liquids, samples or reagents; Photo-bioreactor |
Material | Polypropylene, PTFE, PEEK, Stainless Steel |
Quantity | 1 Ea. |
Thread Size (Metric) | GL 45 |
Autoclavable | Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'