Learn More
Scat™ GL 45 Thread Safety Caps
Block solvent vapors with ventilation during removal with the Scat™ GL 45 Thread Safety Cap. It also features an air valve that combines valve and filter functions and solvents stay clean and components of solvent mixtures cannot evaporate.
Brand:  Scat™ 107019
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10109392
  • 79.79 € / Each
Estimated Shipment
04-12-2018
Add to basket
Description
Description
  • GL 45 screw thread cap
  • Connector 3.2mm O. D.
  • Includes capillaries
Specifications
Specifications
1 Capillary
1 Each
3.2mm
1 Each
Cap
Cap I
Videos