missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Glass Microfiber Filters, Grade MGB
Binder-free glass microfiber filters for analytical and gravimetric analyses and prefiltering.
403.00 € - 5270.00 €
Specifications
Basis Weight | 140 g/m2 |
---|---|
Color | White |
Format | Sheet |
Material | Binder-free Glass Microfiber |
Particle Retention | 1 μm |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15902730
|
Sartorius Stedim Biotech™
FT-2-1102-103257 |
103 x 257 mm |
403.00 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15912730
|
Sartorius Stedim Biotech™
FT-2-1102-210297 |
210 x 297 mm |
995.00 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15922730
|
Sartorius Stedim Biotech™
FT-2-1102-300300 |
300 x 300 mm |
885.00 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15932730
|
Sartorius Stedim Biotech™
FT-2-1102-460570 |
460 x 570 mm |
3120.00 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15942730
|
Sartorius Stedim Biotech™
FT-2-1102-620620 |
620 x 620 mm |
5270.00 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- Binder-free
- Suitable for the following:
- Liquid filtration
- Prefiltration
- Clarification of buffer and reagent solutions
- Analytical sample preparation
- Particle analysis
- Non-sterile, bulk packed
Specifications
140 g/m2 | |
Sheet | |
1 μm | |
210 mL/min. | |
Hydrophilic | |
1 μm |
White | |
Binder-free Glass Microfiber | |
Non-sterile | |
MGB | |
100/Pk. |