missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Glassy Carbon Cylindrical Crucible

Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.

Brand:  Alfa Aesar™ 039008.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15476275

  • 339.00 € / Each


Estimated Shipment date
from Supplier 20-06-2019
Add to basket

Description

Description

Subjected to a high temperature heat treatment which imparts special material properties including significantly improved corrosion resistance and strength.

Glassy carbon offers:
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
Specifications

Specifications

10mL
24mm
Glassy Carbon Crucible
Cylindrical
39mm
Glassy Carbon
Videos
SDS