missing translation for 'onlineSavingsMsg'
Learn More

Gluconic Acid, 50 wt. % Solution in Water, Thermo Scientific Chemicals

Catalog Number 10001020
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
526-95-4, 7732-18-5
C6H12O7
196.155
MFCD00004240
RGHNJXZEOKUKBD-SQOUGZDYSA-N
gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate
10690
CHEBI:33198
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
C(C(C(C(C(C(=O)O)O)O)O)O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
526-95-4, 7732-18-5
C6H12O7
196.155
MFCD00004240
RGHNJXZEOKUKBD-SQOUGZDYSA-N
gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate
10690
CHEBI:33198
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
C(C(C(C(C(C(=O)O)O)O)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White to Yellow
Beilstein 03, 542
Merck Index 15, 4492
Formula Weight 196.16
Percent Purity 49 to 55% (Titrimetry other)
Density 1.23g/mL
Quantity 1 L
Packaging Glass bottle
Physical Form Crystals or Crystalline Powder
Specific Gravity 1.22 to 1.25 (20°C)
Chemical Name or Material Gluconic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'