missing translation for 'onlineSavingsMsg'
Learn More

Glutathione, 98%, for analysis, reduced, Thermo Scientific Chemicals

Catalog Number 10021090
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
70-18-8
C10H17N3O6S
307.32
MFCD00065939
RWSXRVCMGQZWBV-WDSKDSINSA-N
glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
124886
CHEBI:16856
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
70-18-8
C10H17N3O6S
307.32
MFCD00065939
RWSXRVCMGQZWBV-WDSKDSINSA-N
glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
124886
CHEBI:16856
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 182.0°C to 192.0°C
Color White
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Glass bottle
Quantity 5 g
Beilstein 04,II,931
Merck Index 15,4511
Specific Rotation Condition − 16.50 (20.00°C c=2,H2O)
Specific Rotation − 16.50
Solubility Information Solubility in water: very soluble. Other solubilities: freely soluble in dilute alcohol and dimethyl-, formamide, almost insoluble in ethanol
Formula Weight 307.32
Percent Purity 98%
Grade Analysis
Physical Form Crystalline Powder
Chemical Name or Material Glutathione, For analysis, Reduced
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'