missing translation for 'onlineSavingsMsg'
Learn More

Glycine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-4509613
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
623-33-6
C4H9NO2·HCl
139.58
TXTWXQXDMWILOF-UHFFFAOYSA-N
glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor
2723640
ethyl 2-aminoacetate;hydrochloride
CCOC(=O)CN.Cl
This item is not returnable. View return policy
623-33-6
C4H9NO2·HCl
139.58
TXTWXQXDMWILOF-UHFFFAOYSA-N
glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor
2723640
ethyl 2-aminoacetate;hydrochloride
CCOC(=O)CN.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 145°C to 148°C
Color White
pH ≥3 (1% aq. soln.)
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NCH2CO2C2H5·HCl
Quantity 100 g
Beilstein 04,34
Solubility Information Solubility in water: >1000g/L (20°C). Other solubilities: soluble in ethanol
Formula Weight 139.58
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Glycine ethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'