missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
120.00 €
Specifications
| Antigen | IgM (H+L) |
|---|---|
| Concentration | 4.5 mg/mL |
| Applications | Immunoassay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18544524
|
Novus Biologicals
NBP1-75203-0.5mg |
0.5 mg | |||||||
Description
IgM (H+L) Polyclonal specifically detects IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay.Specifications
| IgM (H+L) | |
| Immunoassay | |
| Unconjugated | |
| RUO | |
| Purified Mouse IgM (H&L) | |
| Secondary | |
| Store at 4°C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20°C. Avoid repeated freeze/thaw cycles. |
| 4.5 mg/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| IgM | |
| Affinity purified | |
| Based on IEP, this antibody reacts with heavy mu chains on mouse IgM and light chains on all mouse immunoglobulins |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title