Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, DyLight™ 594, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen 35560
162.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : A0
Additional Details : Weight : 1.81440kg
Description
Product # 35560 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 35560 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 594 Amax= 593 nm; Emax= 618 nm. Mole Dye/Mole Protein Ratio is lot-dependent.
Rabbit IgG (H+L) Polyclonal antibody specifically detects Rabbit IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Rabbit IgG (H+L) | |
Polyclonal | |
DyLight 594 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Rabbit | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.2 | |
Purified Rabbit IgG, whole molecule | |
1 mL | |
Secondary | |
4° C | |
IgG |