Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen 31460
334.40 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : A0
Additional Details : Weight : 1.81440kg
Description
Product # 31460 has been successfully used in Western blot, IHC and IP applications. Product # 31460 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species and with SuperBlock® Blocking Buffers. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 2.0 mL of distilled water (0.8 mg/mL after restoration).
Rabbit IgG (H+L) Polyclonal antibody specifically detects Rabbit IgG (H+L) in Rabbit samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Rabbit IgG (H+L) | |
Polyclonal | |
HRP | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Rabbit | |
Lyophilized |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.8 mg/mL | |
PBS with 15mg/mL BSA, 50mM sucrose and no preservative; pH 7.6 | |
Purified Rabbit IgG, whole molecule | |
2 mL | |
Secondary | |
4° C | |
IgG |