Learn More
Goat anti-Rat IgG (H+L) Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen 31470
219.45 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : A0
Additional Details : Weight : 0.80000kg
Description
This antibody has been successfully used in Western blot, IHC and IP applications. Antibody Specificity: This antibody reacts with the heavy chains of rat IgG and with the light chains common to most rat immunoglobulins, based on immunoelectrophoresis. No antibody was detected against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 2.0 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA
Rat IgG (H+L) Polyclonal antibody specifically detects Rat IgG (H+L) in Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Rat IgG (H+L) | |
Polyclonal | |
HRP | |
Goat | |
2 mL | |
Secondary | |
4° C | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.8 mg/mL | |
PBS with 15mg/mL BSA and no preservative; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Rat | |
Lyophilized |