missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold Germanium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Premion™, 99.9997% (metals basis)

Brand:  Alfa Aesar™ 014172.KJ

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15473495

  • 2050.00 € / Pack of 5

Estimated Shipment date
from Supplier 18-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

Au:Ge; 88:12 wt%
≈2.9g/slug
6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
MFCD02091706
Slug
Premion™
Gold Germanium slug
5 pc.
(metals basis)
Odorless
99.9997%
Videos
SDS