missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold nanorods, 12-18nm dia. x 49-59nm length, dispersion in water, 750nm Absorption, positively charged

Brand:  Alfa Aesar™ 046712.AE

Code : NEW

Additional Details : CAS Number : 7440-57-5 Weight : 0.00010kg

Quantity 100mL

 View more versions of this product

Product Code. 15415035

  • 728.00 € / 100mL


Estimated Shipment date
from Supplier 31-08-2018
Add to basket

Specifications

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
Au
MFCD00003436
23985

Specifications

Gold nanorods
12 to 18nm dia. x 49 to 59nm length, dispersion in water, 750nm Absorption, positively charged
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050
Colorless
100mL
7440-57-5
MFCD00003436
Odorless
gold
23985
Liquid
>60°C (dec.)
Videos
SDS