missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Gore™ STA-PURE™ PFL-Series L/S™ Pump Tubing

High-performance pump tubing for challenging applications

Brand:  Masterflex™ 96242-18

Additional Details : Weight : 0.04990kg

 View more versions of this product

Product Code. 15929076

  • 1359.00 € / Each

Estimated Shipment: 26-01-2023
to see stock.
Add to basket

Description

Description

  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Chemical and pharmaceutical applications
  • Enhanced chemical resistance, including aggressive organic solvents, spallation-free, low gas permeability, opaque white
  • Ideal choice for high purity or long-running, critical pharmaceutical and biopharmaceutical processing applications.
  • The patented composite (perfluoroelastomer) and ePTFE (polytetrafluoroethylene), results in a tube that offers high flex durability for greater resistance to rupture—even at applications up to 60 psi.
  • Long-term pump testing proves that Gore™ STA-PURE™ PFL-Series Pump Tubing lasts hundreds of hours with consistent and repeatable flow rates resulting in one saving time and reducing maintenance by having fewer tubing changes.
  • If one is looking for a flow stable / process stable tubing that withstands CIP (clean-in-place) or SIP (steam-in-place) cleaning—look no further than this tubing!
  • Recommended for use in L/S™ and I/P™ standard and Easy-Load™ pump heads, and L/S™Easy-Load™ II pump heads.
Specifications

Specifications

European Pharmacopoeia; USP; Prop 65 NA (no Prop 65 chemicals)
7.9 mm
1 ft.
Pump Tubing
-40°C
2300 mL/min
Autoclave, SIP
510 mm Hg
Yes
STA-PURE Series PFL
0.35 m
L/S Pump Series
18
3/8 in.
60 psi
150°C
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers