missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 1100 Cellulose Technical Filter Paper
For filtration of coarse particles and gelatinous precipitates.
7.25 € - 29.10 €
Specifications
Material | Cellulose |
---|---|
Particle Retention | 50 to 60 μm |
Pore Size | 50 to 60 μm |
For Use With (Application) | Viscous liquids as Arabic gum, Gelatin, emulsions like Agar-agar, Alginate, protective Paper for filter presses |
Grade | 1100 |
Product Code | Brand | Format | Diameter (Metric) Disc | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Diameter (Metric) Disc | Price | Quantity & Availability | ||||
11759955
|
Fisherbrand™
|
Folded Disc | 125 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11802691
|
Fisherbrand™
|
Disc | 190 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11789945
|
Fisherbrand™
|
Disc | 70 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Creped thin paper
- Wet strengthened
- For filtering viscous liquids as arabic gum, gelatine, emulsions like agar-agar, alginate
Specifications
Cellulose | |
50 to 60 μm | |
1100 | |
0.2 mm |
50 to 60 μm | |
Viscous liquids as Arabic gum, Gelatin, emulsions like Agar-agar, Alginate, protective Paper for filter presses | |
100 Pack |