missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 1101 Cellulose Technical Filter Paper

Used for filtration of fatty products and suitable for hot filtrations.

Brand:  Fisherbrand™

Code : 47

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11759975

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Thick creped paper
  • Wet strengthened
  • To filter syrups, essential oils, varnish, resinous solutions, paints, sweetened juices and wines
Specifications

Specifications

Folded Disc
38 to 75 μm
For syrups, essential oils, varnish, resinous solutions, paints, sweetened juices, wines
100 Pack
150 mm
Cellulose
38 to 75 μm
1101
0.55 mm
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers