missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 1101 Cellulose Technical Filter Paper
Used for filtration of fatty products and suitable for hot filtrations.
Marque: Fisherbrand™
Code : 47
Informations supplémentaires : Weight : 0.78300kg
Description
- Thick creped paper
- Wet strengthened
- To filter syrups, essential oils, varnish, resinous solutions, paints, sweetened juices and wines
Spécification
Folded Disc | |
38 to 75 μm | |
For syrups, essential oils, varnish, resinous solutions, paints, sweetened juices, wines | |
100 Pack | |
250 mm |
Cellulose | |
38 to 75 μm | |
1101 | |
0.55 mm | |
250 mm |