missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 122 General Purpose Filter Papers

Product Code. 11798742 Shop All Fisherbrand Products
Change view
Click to view available options
Format:
Disc
Folded Disc
Diameter (Metric):
70 mm
90 mm
150 mm
190 mm
330 mm
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Diameter (Metric) unitSize
11798742 Folded Disc 190 mm Pack of 100
11748742 Disc 70 mm Pack of 100
11812512 Disc 90 mm Pack of 100
11778742 Folded Disc 150 mm Pack of 100
11728752 Disc 150 mm Pack of 100
11748752 Folded Disc 330 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11798742
Only null left
Add to basket
Edge
Add to basket

A general purpose filter paper used for all general works in the laboratory for excellent and consistent filtration.

  • Very fast filtration velocity
  • Used for general works, hospitals, research laboratories
Applications:
  • Chemical industries, pharmaceutical, cosmetic, food and beverage
  • Specifications

    Format Folded Disc
    Filtration Speed Extremely fast
    For Use With (Application) General works
    Quantity 100/Pk.
    Diameter (Metric) 190 mm
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.