Learn More
Cytiva Whatman™ Grade 1573-1/2 Qualitative Filter Papers
Prepleated to fit conical filter funnels in repetitive or multiple analyses.
Brand:  Cytiva 10314753
Code : 69
Additional Details : Weight : 0.77000kg
Product Code. 11368824
  • 271.00 € / Pack of 100
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
Description
Description
  • Medium-fast filter paper
  • High wet strength
  • Increased total loading capacity
  • Maintained flow rate
Specifications
Specifications
Quantitative Filter Paper
1573-/12
Prepleated
fit conical filter funnels in repetitive or multiple analyses
100/Pk.
320 mm