Learn More
Sartorius Quantitative Grade 1288 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Brand: Sartorius FT-3-206-110
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.Specifications
Disc | |
10 sec. | |
100/Pk. | |
110 mm |
12 to 15 μm | |
1288 | |
0.21 mm |