missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 601 General Purpose Filter Papers

A filter paper used in analytical works in laboratories with medium filtration velocity.

Brand:  Fisherbrand™ 0601A00007

10.58 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.


 View more versions of this product

Product Code. 11576873

  • 17.80 € / Pack of 100
In Stock
Edge
Explore more special offers
Add to basket

Description

Description

  • 73 g/m² weight
  • 0.16mm thick
  • Pore size of 5 to 13μm
  • Filtration velocity of 88 sec
  • Burst strength of 1.95kg/cm²
  • Available in a range of sizes

Specifications

Specifications

Disc
88 seconds
100/Pk.
110 mm
5 to 13 μm
General works, hospitals, research laboratories, schools, chemical industries, cosmetics, food and beverage industries
0.16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.