missing translation for 'onlineSavingsMsg'
Learn More

Advantec™ Grade GF75 Glass Fiber Filters

Product Code. 15712739 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (Metric):
25 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
15712739 25 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15712739 Supplier Advantec™ Supplier No. 0665624
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grade GF75 Glass Fiber Filters for analytical or gravimetric studies.

  • Glass microfiber
  • Highest retention
  • Filters very fine protein precipitates
  • Clarifies chemically aggressive solutions

Specifications

Material Glass Microfiber
Pore Size 0.3 μm
Filter Code GF7525MM
For Use With (Application) High retention filtration of very fine protein precipitates and clarifying chemically aggressive solutions. For analytical or gravimetric studies.
Grade GF75
Quantity 100/Box
Shape Round
Diameter (Metric) 25 mm
Diameter (Metric) Disc 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.