missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Graphite conductive adhesive, alcohol based, Thermo Scientific Chemicals
Click to view available options
Quantity:
30 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.9 |
Boiling Point | 82°C |
Flash Point | 11°C(52°F) |
Ignition Point | 270°C (518°F) |
MDL Number | MFCD01866817 |
Quantity | 30 g |
Vapor Pressure | 48hPa (36mm Hg) at 20°C |
Chemical Name or Material | alcohol based |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'