missing translation for 'onlineSavingsMsg'
Learn More

Graphite foil, 1mm (0.04in) thick, 97% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7058706
Click to view available options
Quantity:
30 x 30 cm
30 x 100 cm
30 x 500 cm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
MFCD00144065
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
MFCD00144065

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 30 x 30 cm
Percent Purity 97%
Physical Form Foil
Assay (metals basis)
Chemical Name or Material Graphite foil, 1mm (0.04 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'