Learn More
Thermo Scientific Chemicals Graphite Fusion Crucible Lid
Brand: Thermo Scientific Chemicals 042315.UA
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Graphite is used for casting and polishing purposes, moulding & Carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
For stock No. 40794 | |
15/Pk | |
Graphite Fusion Crucible Lid |
Graphite | |
2.54cm |
RUO – Research Use Only