missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Graphite Fusion Crucible Lid
143.00 € - 228.00 €
Product Code | Brand | Description | Diameter (Metric) Outer | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Diameter (Metric) Outer | Quantity | Price | Quantity & Availability | |||
15473875
|
Thermo Scientific Chemicals
042315.UA |
For stock No. 40794 | 2.54cm | 15/Pk |
228.00 €
Pack of 15 |
In Stock
Log in to see stock. |
|||
15413895
|
Thermo Scientific Chemicals
043337.UA |
For stock No. 14747 | 3.2cm | 15/Pk. |
143.00 €
Pack of 15 |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||
Description
Graphite is used for casting and polishing purposes, moulding & Carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only