missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Graphite Gas Analysis Crucible

Brand:  Alfa Aesar™ 039809.KN

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15473705

  • 96.86 € / Pack of 100

Estimated Shipment date
from Supplier 18-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

4.75mm
12.6mm
24.2mm
Graphite
9.2mm
For Gas analysis
Graphite Crucible
Videos
SDS