missing translation for 'onlineSavingsMsg'
Learn More

Graphite ink for tantalum capacitors, Thermo Scientific Chemicals

Catalog Number p-7051900
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
MFCD00144065
This item is not returnable. View return policy
MFCD00144065

A graphite ink for use as an undercoat for silver ink in the production of tantalum capacitors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A graphite ink for use as an undercoat for silver ink in the production of tantalum capacitors
TRUSTED_SUSTAINABILITY
Color Black
Odor Ammonia-like
Quantity 250 g
Physical Form Paste
Chemical Name or Material Graphite ink for tantalum capacitors
compliance-icons
Product Identifier
  • Graphite ink for tantalum capacitors
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Graphite , Ammoniumhydroxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'